2-Ethoxy-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

54637-500MG 54637-1G 54637-25G 54637-5G
77329-306EA 300.76 USD
77329-306 77353-154 77387-550 77377-398
2-Ethoxy-4-(trifluoromethyl)benzoic acid ≥97%
2-Ethoxy-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD31642380
CAS Number: 334018-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR