(2-Ethoxy-4-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

54636-500MG 54636-1G 54636-5G
77337-976EA 456.29 USD
77337-976 77353-152 77377-396
(2-Ethoxy-4-(trifluoromethyl)phenyl)methanol ≥97%
(2-Ethoxy-4-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃O₂
MW: 220.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564271
CAS Number: 2368238-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR