2-Ethoxy-4-(trifluoromethyl)benzaldehyde2-Ethoxy-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54635-500MG 54635-1G 54635-5G
77336-820EA 394.49 USD
77336-820 77353-150 77377-394
2-Ethoxy-4-(trifluoromethyl)benzaldehyde2-Ethoxy-4-(trifluoromethyl)benzaldehyde ≥97%
2-Ethoxy-4-(trifluoromethyl)benzaldehyde2-Ethoxy-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18824357
CAS Number: 334018-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR