2-(Methylthio)-4-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54631-500MG 54631-1G 54631-5G
77337-434EA 414.06 USD
77337-434 77353-146 77377-390
2-(Methylthio)-4-(trifluoromethoxy)benzaldehyde ≥97%
2-(Methylthio)-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂S
MW: 236.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757353
CAS Number: 1383825-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR