(4-Methyl-3-nitrophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

54613-1G 54613-5G 54613-25G
77340-732EA 60.77 USD
77340-732 77365-586 77377-380
(4-Methyl-3-nitrophenyl)(morpholino)methanone ≥97%
(4-Methyl-3-nitrophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₄
MW: 250.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01213386
CAS Number: 174482-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR