(4-Methyl-3-nitrophenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

54612-500MG 54612-1G 54612-5G 54612-25G
77329-300EA 176.13 USD
77329-300 77346-530 77365-584 77387-546
(4-Methyl-3-nitrophenyl)(piperidin-1-yl)methanone ≥97%
(4-Methyl-3-nitrophenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₃
MW: 248.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01342166
CAS Number: 349134-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR