N,N-diethyl-4-methyl-3-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

54611-1G 54611-5G 54611-25G
77341-780EA 104.03 USD
77341-780 77365-582 77377-378
N,N-diethyl-4-methyl-3-nitrobenzamide ≥97%
N,N-diethyl-4-methyl-3-nitrobenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₃
MW: 236.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01212994
CAS Number: 330467-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR