N-Cyclopropyl-4-methyl-3-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

54608-1G 54608-5G 54608-25G
77341-776EA 104.03 USD
77341-776 77365-576 77377-372
N-Cyclopropyl-4-methyl-3-nitrobenzamide ≥97%
N-Cyclopropyl-4-methyl-3-nitrobenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01339503
CAS Number: 303133-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR