Methyl 2-ethyl-3,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

54600-500MG 54600-1G 54600-5G
77336-818EA 394.49 USD
77336-818 77353-138 77377-368
Methyl 2-ethyl-3,4-difluorobenzoate ≥97%
Methyl 2-ethyl-3,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22490618
CAS Number: 1824578-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR