Ethyl 2-ethyl-3,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

54599-500MG 54599-1G 54599-5G
77329-294EA 346.08 USD
77329-294 77353-136 77377-366
Ethyl 2-ethyl-3,4-difluorobenzoate ≥97%
Ethyl 2-ethyl-3,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757347
CAS Number: 2706870-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR