(2-Ethyl-3,4-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

54597-500MG 54597-1G 54597-5G
77337-432EA 414.06 USD
77337-432 77353-132 77377-362
(2-Ethyl-3,4-difluorophenyl)methanol ≥97%
(2-Ethyl-3,4-difluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757346
CAS Number: 2710527-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR