2-Ethyl-3,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54596-500MG 54596-1G 54596-5G
77329-290EA 313.12 USD
77329-290 77353-130 77377-360
2-Ethyl-3,4-difluorobenzaldehyde ≥97%
2-Ethyl-3,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD34564268
CAS Number: 2706786-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR