(2-Ethyl-3,4-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54595-500MG 54595-1G 54595-5G
77336-092EA 375.95 USD
77336-092 77353-128 77377-358
(2-Ethyl-3,4-difluorophenyl)(methyl)sulfane ≥97%
(2-Ethyl-3,4-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀F₂S
MW: 188.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564267
CAS Number: 2705615-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR