Ethyl 2-fluoro-4-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

54594-500MG 54594-1G 54594-5G
77336-090EA 375.95 USD
77336-090 77353-126 77377-356
Ethyl 2-fluoro-4-methoxy-3-methylbenzoate ≥97%
Ethyl 2-fluoro-4-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34757345
CAS Number: 2386254-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR