Methyl 2-fluoro-4-methoxy-3-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
54593-500MG
54593-1G
54593-5G
54593-25G
77329-288EA
226.6
USD
77329-288
77353-124
77365-572
77387-538
Methyl 2-fluoro-4-methoxy-3-methylbenzoate ≥97%
Methyl 2-fluoro-4-methoxy-3-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁FO₃ MW: 198.191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34757344 CAS Number: 2384056-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...