(2-Fluoro-4-methoxy-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54590-500MG 54590-1G 54590-5G
77336-816EA 394.49 USD
77336-816 77353-120 77377-352
(2-Fluoro-4-methoxy-3-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-4-methoxy-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
MW: 186.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757342
CAS Number: 2709816-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR