(2-Fluoro-4-methoxy-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

54589-500MG 54589-1G 54589-5G 54589-25G
77329-286EA 238.96 USD
77329-286 77353-118 77365-570 77387-536
(2-Fluoro-4-methoxy-3-methylphenyl)methanol ≥97%
(2-Fluoro-4-methoxy-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757341
CAS Number: 2385645-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR