5-Bromo-3-chloro-4-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

54578-500MG 54578-1G 54578-5G 54578-25G
77329-276EA 130.81 USD
77329-276 77344-346 77365-562 77387-528
5-Bromo-3-chloro-4-iodo-2-methoxypyridine ≥97%
5-Bromo-3-chloro-4-iodo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₄BrClINO
MW: 348.364 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530282
CAS Number: 2734773-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR