Ethyl 2-bromo-5-chloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

54574-1G 54574-5G 54574-25G
77341-524EA 91.67 USD
77341-524 77365-560 77377-342
Ethyl 2-bromo-5-chloroisonicotinate ≥95%
Ethyl 2-bromo-5-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.504 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698305
CAS Number: 1214357-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR