2-Chloro-4-ethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

54563-500MG 54563-1G 54563-5G
77338-114EA 465.56 USD
77338-114 77353-102 77377-338
2-Chloro-4-ethyl-1,1'-biphenyl ≥97%
2-Chloro-4-ethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Cl
MW: 216.706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564264
CAS Number: 2734775-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR