1,2-Dichloro-4-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

54561-500MG 54561-1G 54561-5G
77338-016EA 458.35 USD
77338-016 77353-100 77377-336
1,2-Dichloro-4-ethylbenzene ≥95%
1,2-Dichloro-4-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MW: 175.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092455
CAS Number: 6623-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR