1,3-Dibromo-5-ethyl-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

54551-500MG 54551-1G 54551-5G 54551-25G
77329-274EA 101.97 USD
77329-274 77343-002 77365-558 77377-328
1,3-Dibromo-5-ethyl-2-iodobenzene ≥97%
1,3-Dibromo-5-ethyl-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂I
Storage Temperature: Ambient
MDL Number: MFCD11846042
CAS Number: 1160573-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR