(3-Ethyl-5-(methylthio)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

54544-500MG 54544-1G 54544-5G 54544-25G
77329-272EA 281.19 USD
77329-272 77353-092 77365-556 77387-526
(3-Ethyl-5-(methylthio)phenyl)boronic acid ≥97%
(3-Ethyl-5-(methylthio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₂S
Storage Temperature: Ambient
MDL Number: MFCD34530279
CAS Number: 1115024-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR