4-(1,3-Dioxolan-2-yl)-3-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

54527-500MG 54527-1G 54527-5G 54527-25G
77329-266EA 141.11 USD
77329-266 77344-834 77365-552 77387-522
4-(1,3-Dioxolan-2-yl)-3-(trifluoromethyl)benzoic acid ≥95%
4-(1,3-Dioxolan-2-yl)-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₄
MW: 262.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178718
CAS Number: 2643367-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR