tert-Butyl(4-fluoro-3-(methylthio)phenoxy)dimethylsilane ≥97%

Supplier: AOBChem USA

Warning

54514-500MG 54514-1G 54514-5G 54514-25G
77329-264EA 166.86 USD
77329-264 77346-214 77365-550 77387-520
tert-Butyl(4-fluoro-3-(methylthio)phenoxy)dimethylsilane ≥97%
tert-Butyl(4-fluoro-3-(methylthio)phenoxy)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₃H₂₁FOSSi
MW: 272.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530268
CAS Number: 2734774-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR