(3-Chloro-5-iodophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

54485-500MG 54485-1G 54485-5G 54485-25G
77329-258EA 126.69 USD
77329-258 77344-136 77365-544 77377-318
(3-Chloro-5-iodophenyl)(methyl)sulfane ≥95%
(3-Chloro-5-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₆ClIS
MW: 284.545 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22488872
CAS Number: 1822659-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR