3-((4-Methoxybenzyl)oxy)-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

54477-500MG 54477-1G 54477-5G
77329-254EA 326.51 USD
77329-254 77353-078 77377-316
3-((4-Methoxybenzyl)oxy)-5-(trifluoromethyl)benzoic acid ≥97%
3-((4-Methoxybenzyl)oxy)-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃F₃O₄
MW: 326.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757327
CAS Number: 2807441-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR