1-Bromo-5-iodo-4-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

54466-500MG 54466-1G 54466-5G
77336-204EA 382.13 USD
77336-204 77353-076 77377-314
1-Bromo-5-iodo-4-methoxy-2-methylbenzene ≥97%
1-Bromo-5-iodo-4-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO
Storage Temperature: Ambient
MDL Number: MFCD24115720
CAS Number: 1511204-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR