2-(2-Chloro-6-fluoro-3-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

54463-1G 54463-5G 54463-25G
77341-030EA 66.95 USD
77341-030 77365-540 77377-312
2-(2-Chloro-6-fluoro-3-methylphenyl)propan-2-ol ≥97%
2-(2-Chloro-6-fluoro-3-methylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFO
MW: 202.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530261
CAS Number: 1542853-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR