(6-Isobutoxynaphthalen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

54460-500MG 54460-1G 54460-5G 54460-25G
77329-250EA 225.57 USD
77329-250 77353-072 77365-538 77387-512
(6-Isobutoxynaphthalen-2-yl)methanol ≥97%
(6-Isobutoxynaphthalen-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34757322
CAS Number: 2755723-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR