6-Isobutoxy-2-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

54459-500MG 54459-1G 54459-5G 54459-25G
77329-248EA 189.52 USD
77329-248 77353-070 77365-536 77387-510
6-Isobutoxy-2-naphthaldehyde ≥97%
6-Isobutoxy-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530260
CAS Number: 2734779-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR