Ethyl 6-isobutoxy-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

54458-500MG 54458-1G 54458-5G 54458-25G
77329-246EA 281.19 USD
77329-246 77353-068 77365-534 77387-508
Ethyl 6-isobutoxy-2-naphthoate ≥97%
Ethyl 6-isobutoxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₃
MW: 272.339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757321
CAS Number: 2755720-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR