Methyl 6-isobutoxy-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

54457-500MG 54457-1G 54457-25G 54457-5G
77329-244EA 296.64 USD
77329-244 77353-066 77387-506 77377-308
Methyl 6-isobutoxy-2-naphthoate ≥97%
Methyl 6-isobutoxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₃
MW: 258.312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564260
CAS Number: 2734776-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR