(4-Methoxy-2,3-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

54423-500MG 54423-1G 54423-5G 54423-25G
77329-240EA 120.51 USD
77329-240 77343-748 77365-530 77377-304
(4-Methoxy-2,3-dimethylphenyl)(methyl)sulfane ≥97%
(4-Methoxy-2,3-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28795055
CAS Number: 1803854-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR