Isopropyl 2-bromonicotinate ≥95%

Supplier: AOBChem USA

Warning

54419-500MG 54419-1G 54419-5G 54419-25G
77329-238EA 145.23 USD
77329-238 77345-136 77365-528 77387-502
Isopropyl 2-bromonicotinate ≥95%
Isopropyl 2-bromonicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD34757308
CAS Number: 1695036-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR