(2-Bromo-5-nitrophenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

54376-500MG 54376-1G 54376-5G 54376-25G
77329-236EA 128.75 USD
77329-236 77344-314 77365-524 77377-294
(2-Bromo-5-nitrophenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2-Bromo-5-nitrophenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrN₃O₃
MW: 328.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15466499
CAS Number: 1292135-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR