(2-Bromo-5-nitrophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

54374-500MG 54374-1G 54374-5G 54374-25G
77329-234EA 229.69 USD
77329-234 77353-054 77365-522 77387-500
(2-Bromo-5-nitrophenyl)(morpholino)methanone ≥97%
(2-Bromo-5-nitrophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂O₄
MW: 315.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15466494
CAS Number: 1305414-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR