(Oxybis(4,1-phenylene))dimethanol ≥97%

Supplier: AOBChem USA

Warning

54311-500MG 54311-1G 54311-5G 54311-25G
77329-228EA 190.55 USD
77329-228 77353-050 77365-516 77387-494
(Oxybis(4,1-phenylene))dimethanol ≥97%
(Oxybis(4,1-phenylene))dimethanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₃
MW: 230.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021968
CAS Number: 2350-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR