4-Isobutoxy-2,3-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54309-500MG 54309-1G 54309-5G
77336-876EA 396.55 USD
77336-876 77353-048 77377-292
4-Isobutoxy-2,3-dimethylbenzaldehyde ≥97%
4-Isobutoxy-2,3-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530252
CAS Number: 1523246-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR