2,2-Dichloro-1-(2,6-dimethylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

54297-1G 54297-5G 54297-25G
77341-932EA 108.15 USD
77341-932 77365-514 77377-286
2,2-Dichloro-1-(2,6-dimethylphenyl)ethanol ≥95%
2,2-Dichloro-1-(2,6-dimethylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂O
MW: 219.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530249
CAS Number: 2071258-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR