Methyl 2-fluoro-4-methoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

54265-500MG 54265-1G 54265-5G
77339-338EA 579.89 USD
77339-338 77353-034 77377-278
Methyl 2-fluoro-4-methoxy-5-methylbenzoate ≥97%
Methyl 2-fluoro-4-methoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34757295
CAS Number: 2112658-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR