1-Bromo-3-chloro-2-isopropoxy-5-(trifluoromethyl)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
54253-500MG
54253-1G
54253-5G
54253-25G
77329-224EA
110.21
USD
77329-224
77343-270
77365-510
77377-276
1-Bromo-3-chloro-2-isopropoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-chloro-2-isopropoxy-5-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₁₀H₉BrClF₃O MW: 317.53 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34757287 CAS Number: 2764733-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...