2-(Benzyloxy)-1-bromo-3-chloro-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

54252-500MG 54252-1G 54252-5G 54252-25G
77329-222EA 145.23 USD
77329-222 77345-134 77365-508 77387-490
2-(Benzyloxy)-1-bromo-3-chloro-5-(trifluoromethyl)benzene ≥95%
2-(Benzyloxy)-1-bromo-3-chloro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉BrClF₃O
MW: 365.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757286
CAS Number: 2764730-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR