1-Bromo-3-chloro-2-ethoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

54251-500MG 54251-1G 54251-5G 54251-25G
77329-220EA 133.9 USD
77329-220 77344-414 77365-506 77387-488
1-Bromo-3-chloro-2-ethoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-chloro-2-ethoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD34757285
CAS Number: 2384568-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR