(5-Methoxy-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

54222-500MG 54222-1G 54222-5G 54222-25G
77329-218EA 126.69 USD
77329-218 77344-134 77365-502 77377-272
(5-Methoxy-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥95%
(5-Methoxy-2-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃S
Storage Temperature: Ambient
MDL Number: MFCD34757265
CAS Number: 2639795-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR