2,5-Dichloro-1-methyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

54200-500MG 54200-1G 54200-5G 54200-25G
77329-212EA 120.51 USD
77329-212 77343-746 77365-496 77377-270
2,5-Dichloro-1-methyl-3-nitrobenzene ≥97%
2,5-Dichloro-1-methyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787386
CAS Number: 1807182-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR