4-(Benzyloxy)-5-chloro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

54182-500MG 54182-1G 54182-5G
77335-686EA 363.59 USD
77335-686 77353-028 77377-268
4-(Benzyloxy)-5-chloro-2-methylbenzoic acid ≥97%
4-(Benzyloxy)-5-chloro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34757257
CAS Number: 2409510-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR