1-(Benzyloxy)-4-bromo-2-chloro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

54179-500MG 54179-1G 54179-5G 54179-25G
77329-208EA 177.16 USD
77329-208 77346-600 77365-494 77387-480
1-(Benzyloxy)-4-bromo-2-chloro-5-methylbenzene ≥97%
1-(Benzyloxy)-4-bromo-2-chloro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO
MW: 311.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564253
CAS Number: 2734775-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR