2-Chloro-4-iodo-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

54178-500MG 54178-1G 54178-5G
77329-206EA 307.97 USD
77329-206 77353-024 77377-264
2-Chloro-4-iodo-5-methylphenol ≥97%
2-Chloro-4-iodo-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆ClIO
Storage Temperature: Ambient
MDL Number: MFCD33406175
CAS Number: 786712-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR