5-Chloro-4-ethoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54163-500MG 54163-1G 54163-5G
77338-078EA 462.47 USD
77338-078 77353-020 77377-260
5-Chloro-4-ethoxy-2-methylbenzaldehyde ≥97%
5-Chloro-4-ethoxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246193
CAS Number: 883522-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR